7-Fluoro-2-tetralone ≥98%
Dodavatel: Apollo Scientific
Synonyms:
7-Fluoro-3,4-dihydro-1H-naphthalen-2-one
PC902738-1G
PC902738-250MG
APOSPC902738-1GEA
15480
CZK
APOSPC902738-1G
APOSPC902738-250MG
7-Fluoro-2-tetralone ≥98%
7-Fluoro-2-tetralone
Vzorec:
C₁₀H₉FO MW: 164,176 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04038615 CAS číslo: 29419-15-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...