Tisk…

1-(2-Fluoroethoxy)-4-nitrobenzene 97%

Dodavatel: Apollo Scientific

PC902697-10G PC902697-1G PC902697-5G
APOSPC902697-10GEA 14290 CZK
APOSPC902697-10G APOSPC902697-1G APOSPC902697-5G
1-(2-Fluoroethoxy)-4-nitrobenzene 97%
1-(2-Fluoroethoxy)-4-nitrobenzene
Vzorec: C₈H₈FNO₃
MW: 185.152 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09743783
CAS číslo: 109230-65-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR