Tisk…

Ethyl 2-fluoro-3-oxopentanoate 95%

Dodavatel: Apollo Scientific

PC902638-1G PC902638-5G
APOSPC902638-1GEA 2900 CZK
APOSPC902638-1G APOSPC902638-5G
Ethyl 2-fluoro-3-oxopentanoate 95%
Ethyl 2-fluoro-3-oxopentanoate
Vzorec: C₇H₁₁FO₃
MW: 162,159 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09753153
CAS číslo: 759-67-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR