Tisk…

N-Acetyl 5-fluoro-2-methylaniline 98%

Dodavatel: Apollo Scientific

Synonyms: 2-Acetamido-4-fluorotoluene

PC902344-25G
APOSPC902344-25GEA 3970 CZK
APOSPC902344-25G
N-Acetyl 5-fluoro-2-methylaniline 98%
N-Acetyl 5-fluoro-2-methylaniline
Vzorec: C₉H₁₀FNO
MW: 167,18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00045064
CAS číslo: 366-49-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR