Tisk…

6-Fluoro-1,3-benzoxazole-2-thiol 98%

Dodavatel: Apollo Scientific

PC902320-1G PC902320-5G
APOSPC902320-1GEA 7670 CZK
APOSPC902320-1G APOSPC902320-5G
6-Fluoro-1,3-benzoxazole-2-thiol 98%
6-Fluoro-1,3-benzoxazole-2-thiol
Vzorec: C₇H₄FNOS
Storage Temperature: Ambient
MDL Number: MFCD18861560
CAS číslo: 145096-57-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR