Tisk…

1-Ethyl-6,7-difluorobenzimidazole 98%

Dodavatel: Apollo Scientific

PC902255-100G PC902255-5G PC902255-25G
APOSPC902255-100GEA 17990 CZK
APOSPC902255-100G APOSPC902255-5G APOSPC902255-25G
1-Ethyl-6,7-difluorobenzimidazole 98%
1-Ethyl-6,7-difluorobenzimidazole
Vzorec: C₉H₈F₂N₂
MW: 182,17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19684115
CAS číslo: 1314987-78-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR