2-Methyl-4-(trifluoromethyl)benzonitrile 98%
Dodavatel: Apollo Scientific
PC902122-1G
PC902122-250MG
APOSPC902122-1GEA
19110
CZK
APOSPC902122-1G
APOSPC902122-250MG
2-Methyl-4-(trifluoromethyl)benzonitrile 98%
2-Methyl-4-(trifluoromethyl)benzonitrile
Vzorec:
C₉H₆F₃N MW: 185,146 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD13185724 CAS číslo: 362640-56-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...