4-Chloro-2,6-difluoronitrobenzene 95%
Dodavatel: Apollo Scientific
PC902118-1G
PC902118-5G
APOSPC902118-1GEA
1540
CZK
APOSPC902118-1G
APOSPC902118-5G
4-Chloro-2,6-difluoronitrobenzene 95%
4-Chloro-2,6-difluoronitrobenzene
Vzorec:
C₆H₂ClF₂NO₂ Storage Temperature: Ambient |
MDL Number:
MFCD20039793 CAS číslo: 136272-31-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...