4-Fluoro-N-methyl-3-nitrobenzamide 95%
Dodavatel: Apollo Scientific
PC902104-1G
PC902104-5G
APOSPC902104-1GEA
1160
CZK
APOSPC902104-1G
APOSPC902104-5G
4-Fluoro-N-methyl-3-nitrobenzamide 95%
4-Fluoro-N-methyl-3-nitrobenzamide
Vzorec:
C₈H₇FN₂O₃ Storage Temperature: Ambient |
MDL Number:
MFCD11855197 CAS číslo: 475216-25-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...