Tisk…

N-[(2-Fluorophenyl)methyl]cyclopropanamine 97%

Dodavatel: Apollo Scientific

PC902081-1G PC902081-5G
APOSPC902081-1GEA 6220 CZK
APOSPC902081-1G APOSPC902081-5G
N-[(2-Fluorophenyl)methyl]cyclopropanamine 97%
N-[(2-Fluorophenyl)methyl]cyclopropanamine
Vzorec: C₁₀H₁₂FN
MW: 165.207 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04035532
CAS číslo: 625435-02-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR