Tisk…

5-Amino-3-(2-fluorophenyl)-1-methylpyrazole 96%

Dodavatel: Apollo Scientific

PC902069-1G PC902069-5G
APOSPC902069-1GEA 6220 CZK
APOSPC902069-1G APOSPC902069-5G
5-Amino-3-(2-fluorophenyl)-1-methylpyrazole 96%
5-Amino-3-(2-fluorophenyl)-1-methylpyrazole
Vzorec: C₁₀H₁₀FN₃
MW: 191,205 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11209693
CAS číslo: 1012879-56-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR