Tisk…

2-(3-Fluorophenyl)-3-oxobutanenitrile 96%

Dodavatel: Apollo Scientific

PC902055-1G PC902055-5G
APOSPC902055-1GEA 6220 CZK
APOSPC902055-1G APOSPC902055-5G
2-(3-Fluorophenyl)-3-oxobutanenitrile 96%
2-(3-Fluorophenyl)-3-oxobutanenitrile
Vzorec: C₁₀H₈FNO
MW: 177,175 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11208702
CAS číslo: 446-74-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR