5-Bromo-1,2-difluoro-3-nitrobenzene 95%
Dodavatel: Apollo Scientific
PC901988-5G
APOSPC901988-5GEA
2150
CZK
APOSPC901988-5G
5-Bromo-1,2-difluoro-3-nitrobenzene 95%
5-Bromo-1,2-difluoro-3-nitrobenzene
Vzorec:
C₆H₂BrF₂NO₂ MW: 237,986 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18311821 CAS číslo: 1261988-16-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...