Tisk…

4,6-Difluoro-2-methyl-1H-benzo[d]imidazole 98%

Dodavatel: Apollo Scientific

PC901967-1G PC901967-5G
APOSPC901967-1GEA 6220 CZK
APOSPC901967-1G APOSPC901967-5G
4,6-Difluoro-2-methyl-1H-benzo[d]imidazole 98%
4,6-Difluoro-2-methyl-1H-benzo[d]imidazole
Vzorec: C₈H₆F₂N₂
Storage Temperature: Ambient
MDL Number: MFCD09859884
CAS číslo: 874814-18-3

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR