Tisk…

5-Fluorobenzo[d]thiazole ≥98%

Dodavatel: Apollo Scientific

PC901862-100MG PC901862-1G PC901862-250MG
APOSPC901862-100MGEA 2440 CZK
APOSPC901862-100MG APOSPC901862-1G APOSPC901862-250MG
5-Fluorobenzo[d]thiazole ≥98%
5-Fluorobenzo[d]thiazole
Vzorec: C₇H₄FNS
MW: 153,177 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06659658
CAS číslo: 1644-85-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR