Tisk…

2-(4-Fluorophenyl)-2-methylpropanenitrile 98%

Dodavatel: Apollo Scientific

PC901813-1G
APOSPC901813-1GEA 12200 CZK
APOSPC901813-1G
2-(4-Fluorophenyl)-2-methylpropanenitrile 98%
2-(4-Fluorophenyl)-2-methylpropanenitrile
Vzorec: C₁₀H₁₀FN
MW: 163,192 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11036570
CAS číslo: 93748-09-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR