2-Fluoro-6-methoxy-3-methylphenylboronic acid 98%
Dodavatel: Apollo Scientific
Synonyms:
(2-Fluoro-6-methoxy-3-methylphenyl)boronic acid
PC901650-1G
PC901650-5G
APOSPC901650-1GEA
3540
CZK
APOSPC901650-1G
APOSPC901650-5G
2-Fluoro-6-methoxy-3-methylphenylboronic acid 98%
2-Fluoro-6-methoxy-3-methylphenylboronic acid
Vzorec:
C₈H₁₀BFO₃ MW: 183,973 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11856029 CAS číslo: 1451392-12-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...