Tisk…

3-Methyl-4-(trifluoromethyl)phenylboronic acid 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: 3-Methyl-4-(trifluoromethyl)benzeneboronic acid, (3-Methyl-4-(trifluoromethyl)phenyl)boronic acid

PC901640-1G PC901640-5G
APOSPC901640-1GEA 3380 CZK
APOSPC901640-1G APOSPC901640-5G
3-Methyl-4-(trifluoromethyl)phenylboronic acid 98%
3-Methyl-4-(trifluoromethyl)phenylboronic acid
Vzorec: C₈H₈BF₃O₂
MW: 203,954 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08706260
CAS číslo: 864759-67-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR