Tisk…
Methyl 3-(3-fluorophenyl)propanoate 98%
Dodavatel: Apollo Scientific
PC901617-1G PC901617-5G
APOSPC901617-1GEA 4400 CZK
APOSPC901617-1G APOSPC901617-5G
Methyl 3-(3-fluorophenyl)propanoate 98%
Methyl 3-(3-fluorophenyl)propanoate
Vzorec: C₁₀H₁₁FO₂
MW: 182,192 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12031749
CAS číslo: 425704-52-5