Tisk…

3-Bromo-2-fluoro-5-methylbenzaldehyde 95%

Dodavatel: Apollo Scientific
Varování

PC901523-1G PC901523-5G
APOSPC901523-1GEA 2480 CZK
APOSPC901523-1G APOSPC901523-5G
3-Bromo-2-fluoro-5-methylbenzaldehyde 95%
3-Bromo-2-fluoro-5-methylbenzaldehyde
Vzorec: C₈H₆BrFO
MW: 217,035 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12756718
CAS číslo: 1257665-03-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR