Tisk…
1-Methyl-3-trifluoromethylpyrazole-4-boronic acid 95%
Dodavatel: Apollo Scientific
Synonyms: (1-Methyl-3-(trifluoromethyl)-1H-pyrazol-4-yl)boronic acid
PC901442-1G
APOSPC901442-1GEA 3310 CZK
APOSPC901442-1G
1-Methyl-3-trifluoromethylpyrazole-4-boronic acid 95%
1-Methyl-3-trifluoromethylpyrazole-4-boronic acid
Vzorec: C₅H₆BF₃N₂O₂
MW: 193,92 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13619875
CAS číslo: 1138450-30-2