Tisk…

3,5-Difluoro-4-methylphenylboronic acid 95%

Dodavatel: Apollo Scientific
Varování

PC901273-1G PC901273-250MG
APOSPC901273-1GEA 4370 CZK
APOSPC901273-1G APOSPC901273-250MG
3,5-Difluoro-4-methylphenylboronic acid 95%
3,5-Difluoro-4-methylphenylboronic acid
Vzorec: C₇H₇BF₂O₂
MW: 171,937 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21365108
CAS číslo: 1621332-09-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR