(4-Chloro-2-fluoro-3-methylphenyl)boronic acid 97%
Dodavatel: Apollo Scientific
Synonyms:
4-Chloro-2-fluoro-3-methylphenylboronic acid
PC901263-1G
PC901263-5G
APOSPC901263-1GEA
5060
CZK
APOSPC901263-1G
APOSPC901263-5G
(4-Chloro-2-fluoro-3-methylphenyl)boronic acid 97%
(4-Chloro-2-fluoro-3-methylphenyl)boronic acid
Vzorec:
C₇H₇BClFO₂ MW: 188.392 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18447609 CAS číslo: 944128-92-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...