Tisk…

2-Ethoxy-4-fluoro-5-methylphenylboronic acid 97%

Dodavatel: Apollo Scientific

PC900901-10G PC900901-1G PC900901-5G
APOSPC900901-10GEA 21490 CZK
APOSPC900901-10G APOSPC900901-1G APOSPC900901-5G
2-Ethoxy-4-fluoro-5-methylphenylboronic acid 97%
2-Ethoxy-4-fluoro-5-methylphenylboronic acid
Vzorec: C₉H₁₂BFO₃
MW: 197,999 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19684096
CAS číslo: 2096337-59-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR