Tisk…

5-Fluoro-2-isopropoxypyridine 98%

Dodavatel: Apollo Scientific

PC900586-1G PC900586-5G
APOSPC900586-1GEA 6220 CZK
APOSPC900586-1G APOSPC900586-5G
5-Fluoro-2-isopropoxypyridine 98%
5-Fluoro-2-isopropoxypyridine
Vzorec: C₈H₁₀FNO
MW: 155.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18914533
CAS číslo: 1305322-99-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR