Methyl 2-(4-amino-2-fluorophenyl)acetate 98%
Dodavatel: Apollo Scientific
PC900415-10G
PC900415-1G
PC900415-5G
APOSPC900415-10GEA
26450
CZK
APOSPC900415-10G
APOSPC900415-1G
APOSPC900415-5G
Methyl 2-(4-amino-2-fluorophenyl)acetate 98%
Methyl 2-(4-amino-2-fluorophenyl)acetate
Vzorec:
C₉H₁₀FNO₂ MW: 183,18 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD20727501 CAS číslo: 192650-54-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...