Tisk…

2,3-Difluoro-N-methylbenzamide 95%

Dodavatel: Apollo Scientific

PC900235-1G PC900235-5G
APOSPC900235-1GEA 7050 CZK
APOSPC900235-1G APOSPC900235-5G
2,3-Difluoro-N-methylbenzamide 95%
2,3-Difluoro-N-methylbenzamide
Vzorec: C₈H₇F₂NO
Storage Temperature: Ambient
MDL Number: MFCD16836083
CAS číslo: 1250397-72-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR