Tisk…

5-Fluoro-2-(2-methylpropoxy)benzonitrile 98%

Dodavatel: Apollo Scientific

PC900220-1G PC900220-5G
APOSPC900220-1GEA 6220 CZK
APOSPC900220-1G APOSPC900220-5G
5-Fluoro-2-(2-methylpropoxy)benzonitrile 98%
5-Fluoro-2-(2-methylpropoxy)benzonitrile
Vzorec: C₁₁H₁₂FNO
MW: 193,217 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18860665
CAS číslo: 1340270-09-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR