Tisk…

2,3-Difluoro-1-methyl-4-nitrobenzene 98%

Dodavatel: Apollo Scientific

PC900212-25G
APOSPC900212-25GEA 5390 CZK
APOSPC900212-25G
2,3-Difluoro-1-methyl-4-nitrobenzene 98%
2,3-Difluoro-1-methyl-4-nitrobenzene
Vzorec: C₇H₅F₂NO₂
MW: 173,117 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21603708
CAS číslo: 932373-72-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR