Tisk…

b-Amino-2-fluorobenzeneethanol 97%

Dodavatel: Apollo Scientific

PC900196-1G PC900196-250MG
APOSPC900196-1GEA 17590 CZK
APOSPC900196-1G APOSPC900196-250MG
b-Amino-2-fluorobenzeneethanol 97%
b-Amino-2-fluorobenzeneethanol
Vzorec: C₈H₁₀FNO
MW: 155,17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09759236
CAS číslo: 179811-62-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR