(S)-b-Amino-2-fluorobenzenepropanol 97%
Dodavatel: Apollo Scientific
PC900186-1G
PC900186-250MG
PC900186-500MG
APOSPC900186-1GEA
24800
CZK
APOSPC900186-1G
APOSPC900186-250MG
APOSPC900186-500MG
(S)-b-Amino-2-fluorobenzenepropanol 97%
(S)-b-Amino-2-fluorobenzenepropanol
Vzorec:
C₉H₁₂FNO Storage Temperature: Ambient |
MDL Number:
MFCD17214632 CAS číslo: 148021-84-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...