Tisk…

N-[2-(2-Fluorophenyl)propan-2-yl]acetamide 97%

Dodavatel: Apollo Scientific

PC900146-10G PC900146-1G PC900146-5G
APOSPC900146-10GEA 21490 CZK
APOSPC900146-10G APOSPC900146-1G APOSPC900146-5G
N-[2-(2-Fluorophenyl)propan-2-yl]acetamide 97%
N-[2-(2-Fluorophenyl)propan-2-yl]acetamide
Vzorec: C₁₁H₁₄FNO
Storage Temperature: Ambient
MDL Number: MFCD22689116
CAS číslo: 1420800-24-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR