1-Fluoro-4-methyl-2-propoxybenzene 98%
Dodavatel: Apollo Scientific
PC900062-1G
PC900062-25G
PC900062-5G
APOSPC900062-1GEA
3110
CZK
APOSPC900062-1G
APOSPC900062-25G
APOSPC900062-5G
1-Fluoro-4-methyl-2-propoxybenzene 98%
1-Fluoro-4-methyl-2-propoxybenzene
Vzorec:
C₁₀H₁₃FO Storage Temperature: Ambient |
MDL Number:
MFCD19237203 CAS číslo: 1311197-87-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...