1-Bromo-3,4-difluoro-2-methoxybenzene 97%
Dodavatel: Apollo Scientific
PC900061-1G
PC900061-5G
APOSPC900061-1GEA
1780
CZK
APOSPC900061-1G
APOSPC900061-5G
1-Bromo-3,4-difluoro-2-methoxybenzene 97%
1-Bromo-3,4-difluoro-2-methoxybenzene
Vzorec:
C₇H₅BrF₂O MW: 223,015 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD16295239 CAS číslo: 888318-22-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...