Tisk…
Methyl 4-ethoxy-2-fluorobenzoate 98%
Dodavatel: Apollo Scientific
PC900056-1G PC900056-5G
APOSPC900056-1GEA 4140 CZK
APOSPC900056-1G APOSPC900056-5G
Methyl 4-ethoxy-2-fluorobenzoate 98%
Methyl 4-ethoxy-2-fluorobenzoate
Vzorec: C₁₀H₁₁FO₃
MW: 198.191 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19684122
CAS číslo: 1314987-36-4