Tisk…

[2-Methyl-6-(trifluoromethyl)pyridin-3-yl]methanol

Dodavatel: Apollo Scientific

PC8983-1G PC8983-5G
APOSPC8983-1GEA 3110 CZK
APOSPC8983-1G APOSPC8983-5G
[2-Methyl-6-(trifluoromethyl)pyridin-3-yl]methanol
[2-Methyl-6-(trifluoromethyl)pyridin-3-yl]methanol
Vzorec: C₈H₈F₃NO
MW: 191,15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03407989
CAS číslo: 681260-50-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR