Tisk…

2,2,3,3-Tetrafluoropropanoic anhydride

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 3H-Perfluoropropanoic anhydride, 2,2,3,3-Tetrafluoropropionic anhydride

PC8916-1G PC8916-5G
APOSPC8916-1GEA 5200 CZK
APOSPC8916-1G APOSPC8916-5G
2,2,3,3-Tetrafluoropropanoic anhydride
2,2,3,3-Tetrafluoropropanoic anhydride
Vzorec: C₆H₂F₈O₃
MW: 274,066 g/mol
Bod varu: 65…70 °C (80 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD17010235
CAS číslo: 337-83-7
UN: 3265
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR