Tisk…

2-Methyl-4-oxo-4-[4-(trifluoromethyl)phenyl]butanoic acid

Dodavatel: Apollo Scientific

PC8906-1G PC8906-5G
APOSPC8906-1GEA 7710 CZK
APOSPC8906-1G APOSPC8906-5G
2-Methyl-4-oxo-4-[4-(trifluoromethyl)phenyl]butanoic acid
2-Methyl-4-oxo-4-[4-(trifluoromethyl)phenyl]butanoic acid
Vzorec: C₁₂H₁₁F₃O₃
MW: 260,209 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04972252
CAS číslo: 75380-98-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR