1-Bromo-2-chloro-4,5-difluorobenzene 97%
Dodavatel: Apollo Scientific
|
Varování
|
PC8715-1G
PC8715-5G
PC8715-250MG
APOSPC8715-1GEA
2900
CZK
APOSPC8715-1G
APOSPC8715-5G
APOSPC8715-250MG
1-Bromo-2-chloro-4,5-difluorobenzene 97%
1-Bromo-2-chloro-4,5-difluorobenzene
Vzorec:
C₆H₂BrClF₂ Storage Temperature: Ambient |
MDL Number:
MFCD00142586 CAS číslo: 59447-06-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...