Tisk…

2-Chloro-N-ethyl-6-fluorobenzylamine

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: N-(2-Chloro-6-fluorobenzyl)ethylamine, N-(2-Chloro-6-fluorobenzyl)ethanamine

PC8700-1G PC8700-250MG
APOSPC8700-1GEA 3480 CZK
APOSPC8700-1G APOSPC8700-250MG
2-Chloro-N-ethyl-6-fluorobenzylamine
2-Chloro-N-ethyl-6-fluorobenzylamine
Vzorec: C₉H₁₁ClFN
Storage Temperature: Ambient
MDL Number: MFCD00053426
CAS číslo: 62924-59-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR