3-Fluoro-4-propoxybenzeneboronic acid
Dodavatel: Apollo Scientific
|
Varování
|
PC8650-1G
PC8650-5G
PC8650-25G
APOSPC8650-1GEA
622
CZK
APOSPC8650-1G
APOSPC8650-5G
APOSPC8650-25G
3-Fluoro-4-propoxybenzeneboronic acid
3-Fluoro-4-propoxybenzeneboronic acid
Vzorec:
C₉H₁₂BFO₃ Storage Temperature: Refrigerator |
MDL Number:
MFCD06411285 CAS číslo: 192376-68-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...