Tisk…

Pentafluorophenyl 6-thien-2-ylnicotinate 97%

Dodavatel: Apollo Scientific

Synonyms: Pentafluorophenyl 2-thien-2-ylpyridine-5-carboxylate

PC8536-1G PC8536-250MG
APOSPC8536-1GEA 6250 CZK
APOSPC8536-1G APOSPC8536-250MG
Pentafluorophenyl 6-thien-2-ylnicotinate 97%
Pentafluorophenyl 6-thien-2-ylnicotinate
Vzorec: C₁₆H₆F₅NO₂S
MW: 371.281 g/mol
Bod tání: 139.5…141.5 °C
Storage Temperature: Ambient
MDL Number: MFCD09702374
CAS číslo: 926921-59-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR