Pentafluorophenyl 2-thiomorpholin-4-ylisonicotinate 97%
Dodavatel: Apollo Scientific
Synonyms:
Pentafluorophenyl 2-thiomorpholin-4-ylpyridine-4-carboxylate
PC8532-1G
PC8532-250MG
APOSPC8532-1GEA
5660
CZK
APOSPC8532-1G
APOSPC8532-250MG
Pentafluorophenyl 2-thiomorpholin-4-ylisonicotinate 97%
Pentafluorophenyl 2-thiomorpholin-4-ylisonicotinate
Vzorec:
C₁₆H₁₁F₅N₂O₂S MW: 390,328 g/mol Bod tání: 11…100,5 °C Storage Temperature: Ambient |
MDL Number:
MFCD09702363 CAS číslo: 934570-42-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...