Tisk…

Pentafluorophenyl 6-phenylnicotinate 97%

Dodavatel: Apollo Scientific

Synonyms: Pentafluorophenyl 2-phenylpyridine-5-carboxylate

PC8530-1G PC8530-250MG
APOSPC8530-1GEA 7280 CZK
APOSPC8530-1G APOSPC8530-250MG
Pentafluorophenyl 6-phenylnicotinate 97%
Pentafluorophenyl 6-phenylnicotinate
Vzorec: C₁₈H₈F₅NO₂
MW: 365.254 g/mol
Bod tání: 113…115 °C
Storage Temperature: Ambient
MDL Number: MFCD09702345
CAS číslo: 934570-41-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR