Pentafluorophenyl 6-(tetrahydropyran-4-yloxy)nicotinate 97%
Dodavatel: Apollo Scientific
Synonyms:
Pentafluorophenyl-2-(tetrahydropyran-4-yloxy)pyridine-5-carboxylate
PC8512-1G
PC8512-250MG
APOSPC8512-1GEA
5660
CZK
APOSPC8512-1G
APOSPC8512-250MG
Pentafluorophenyl 6-(tetrahydropyran-4-yloxy)nicotinate 97%
Pentafluorophenyl 6-(tetrahydropyran-4-yloxy)nicotinate
Vzorec:
C₁₇H₁₂F₅NO₄ MW: 389,273 g/mol Bod tání: 114,5…118 °C Storage Temperature: Ambient |
MDL Number:
MFCD09702389 CAS číslo: 910036-96-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...