Tisk…

3-(Trifluoromethylthio)phenacyl bromide

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 2-Bromo-3'-(trifluoromethylthio)acetophenone

PC8507-1G PC8507-5G
APOSPC8507-1GEA 7940 CZK
APOSPC8507-1G APOSPC8507-5G
3-(Trifluoromethylthio)phenacyl bromide
3-(Trifluoromethylthio)phenacyl bromide
Vzorec: C₉H₆BrF₃OS
MW: 299,108 g/mol
Bod tání: 32…34 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03094515
CAS číslo: 56773-34-3
UN: 3261
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR