Tisk…

2-(Aminomethyl)-3-(trifluoromethyl)pyridine hydrochloride

Dodavatel: Apollo Scientific
Varování

PC8370-250MG PC8370-1G
APOSPC8370-250MGEA 3540 CZK
APOSPC8370-250MG APOSPC8370-1G
2-(Aminomethyl)-3-(trifluoromethyl)pyridine hydrochloride
2-(Aminomethyl)-3-(trifluoromethyl)pyridine hydrochloride
Vzorec: C₇H₇F₃N₂·ClH
Storage Temperature: Refrigerator
MDL Number: MFCD04117764
CAS číslo: 1187932-68-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR