Tisk…

3-Fluoro-2-methoxy-5-(methylthio)benzeneboronic acid 98%

Dodavatel: Apollo Scientific
Varování

PC8339-1G PC8339-250MG PC8339-5G
APOSPC8339-1GEA 8870 CZK
APOSPC8339-1G APOSPC8339-250MG APOSPC8339-5G
3-Fluoro-2-methoxy-5-(methylthio)benzeneboronic acid 98%
3-Fluoro-2-methoxy-5-(methylthio)benzeneboronic acid
Vzorec: C₈H₁₀BFO₃S
MW: 216,038 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09878322
CAS číslo: 957065-83-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR