Tisk…

2,6-Difluoro-4-methoxybenzamide 98%

Dodavatel: Apollo Scientific

PC8315-10G PC8315-5G PC8315-1G
APOSPC8315-10GEA 8100 CZK
APOSPC8315-10G APOSPC8315-5G APOSPC8315-1G
2,6-Difluoro-4-methoxybenzamide 98%
2,6-Difluoro-4-methoxybenzamide
Vzorec: C₈H₇F₂NO₂
MW: 187,143 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03094497
CAS číslo: 125369-57-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR