Tisk…

4-Amino-2,6-difluorophenol 95%

Dodavatel: Apollo Scientific
Varování

Synonyms: 3,5-Difluoro-4-hydroxyaniline

PC8311-1G PC8311-5G
APOSPC8311-1GEA 3740 CZK
APOSPC8311-1G APOSPC8311-5G
4-Amino-2,6-difluorophenol 95%
4-Amino-2,6-difluorophenol
Vzorec: C₆H₅F₂NO
MW: 145.107 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03094493
CAS číslo: 126058-97-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR